MF-Millipore™ Membrane Filter, 0.45 μm
MF-Millipore™ Membrane Filter, 0.45 μm
Actual product may vary
Fisherbrand
MF-Millipore™ Membrane Filter, 0.45 μm
MF-Millipore™ Membrane Filter are mixed cellulose esters (MCE) membrane, hydrophilic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)DimensionsLxWColorNo. per Case
FKS1052920225 mmWhite100 Pk.
FKS1003441047 mmWhite100 Pk.
Catalog number FKS10529202
Price (SGD)
788.00
PK
Add to cart
Diameter (Metric):
25 mm
Color:
White
No. per Case:
100 Pk.
Price (SGD)
788.00
PK
Add to cart
Designed for a wide range of analytical and research applications. MF-Millipore™ Mixed Cellulose Ester Membranes, 0.45μm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features and Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
AutoclavableNot Autoclavable
Bubble Point≥1.8 bar, air with water
ColorWhite
DisposableReusable
Filter TypeMembrane Filter
FormatDisc
MaterialMCE
Pore Size0.45 μm
SterilityNon-sterile
Flow Rate60 mL/min. x cm2
Gas Flow Rate4 L/min./cm2
HydrophobicityHydrophilic
Membrane TypePlain
No. per Case100 Pk.
Quantity100 Pk.
Surface TypePlain
Thickness (Metric)150 μm
TypeFilter Membranes
WettabilityHydrophilic
Diameter (English)1 in.
Diameter (Metric)25 mm
Unit SizePK