Whatman™ Polycap Disposable Capsules, 36 AS
Whatman™ Polycap Disposable Capsules, 36 AS
Actual product may vary
Fisherbrand
Whatman™ Polycap Disposable Capsules, 36 AS
Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.
Have Questions?
Catalog NumberPore SizeInlet StyleOutlet StyleSterility
FKS105271110.2 μmSB inletSB outletSterile
Catalog number FKS10527111
Price (SGD)
285.00
EA
Add to cart
Pore Size:
0.2 μm
Inlet Style:
SB inlet
Outlet Style:
SB outlet
Sterility:
Sterile
Price (SGD)
285.00
EA
Add to cart
Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.
  • First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
  • Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
  • Housing thermally fused (no glues, adhesives, or extraneous materials).
  • Integrity-testable by bubble point, pressure decay, or forward flow methods.
  • Provides highly effective filtration area in a small size.
  • Autoclavable; some presterilized with gamma irradiation.
  • Manufactured in clean room facilities under ISO Quality Systems.
Specifications
AutoclavableAutoclavable
DescriptionPolycap AS 36 Capsule Filter, sterile, 0.2 μm, SB inlet and outlet (1 pc)
Material (Membrane)Nylon
Pore Size0.2 μm
SterilitySterile
Filtration Area400 cm2
For Use With (Application)Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Housing MaterialPolypropylene
Inlet StyleSB inlet
Max. Pressure4.1 bar (60 psi)
Outlet StyleSB outlet
Unit SizeEA