Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Actual product may vary
Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Fisherbrand
Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Have Questions?
Catalog NumberDimensions (L x W)
FKS1051777120 x 20 cm
Catalog number FKS10517771
Price (SGD)
602.00
PK
Add to cart
Dimensions (L x W):
20 x 20 cm
Price (SGD)
602.00
PK
Add to cart
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Alugram™ Aluminum Sheets for TLC
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
Backing MaterialAlugram™ Aluminum
MaterialSilica 60
Particle Size5 to 17 μm
Pore Size60 Å
Quantity25/Pk.
TypeTLC Plate
Fluorescent IndicatorUV254
Pore Volume0.75 mL/g
SorbentSilica 60
Specific Surface∼500 m2/g
Thickness (Metric)0.2 mm
Dimensions (L x W)20 x 20 cm
Unit SizePK