Nunc™ Disposable Plastic Centrifuge Tubes
Nunc™ Disposable Plastic Centrifuge Tubes
Actual product may vary
Fisherbrand
Nunc™ Disposable Plastic Centrifuge Tubes
Maintain a high level of accuracy while increasing lab safety with these disposable plastic centrifuge tubes, which are an excellent alternative to glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Bottom ShapeSterility
FKS1051706114 mLConicalSterile
FIS12-565-2395 mLRoundNon-sterile
Catalog number FKS10517061
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
14 mL
Bottom Shape:
Conical
Sterility:
Sterile
Maintain a high level of accuracy while increasing lab safety with Thermo Scientific™ Nunc™ Disposable Plastic Centrifuge Tubes, which are an excellent alternative to glass. Selected tubes are relative centrifugal force (RCF) tested. Nunc Labware Products are made from high purity resins and molded using our state of the art processes.
Features:
  • Polyethylene (PE) and polypropylene (PP) tubes are particularly useful in serology where the low adsorption of proteins is of great importance
  • Polystyrene (PS) tubes are transparent and suitable for a broad range of applications
  • Polyethylene and polypropylene tubes have good chemical resistance and high mechanical strength
  • Different cap types are available for a variety of applications and fit most standard centrifuges
Includes:
Cat. nos. 343141, 345608 supplied with mounted stoppers for sterile work.
Specifications
Capacity (Metric)14 mL
DisposableSingle-use
MaterialPolystyrene
No. per Case500
SterilitySterile
ShapeConical
Bottom ShapeConical
Graduations5 and 10 mL
Product TypeCentrifuge Tube
Max. RCF3000 x g
Unit SizePK