Natural Rubber Double Bellows
Natural Rubber Double Bellows
Actual product may vary
Natural Rubber Double Bellows
Natural Rubber Double Bellows
Fisherbrand
Natural Rubber Double Bellows
Use this natural rubber bellows for a variety of applications. Deutsch Neumann™ Natural Rubber Double Bellows feature inlet and outlet valves and a short piece of 3mm bore tubing connected to the outlet.
Have Questions?
Catalog number FKS10507951
Price (SGD)
147.00
EA
Add to cart
Price (SGD)
147.00
EA
Add to cart
Use this natural rubber bellows for a variety of applications. Deutsch Neumann™ Natural Rubber Double Bellows feature inlet and outlet valves and a short piece of 3mm bore tubing connected to the outlet.
  • Double spray bellows  with net
  • Manufactured from black natural rubber
Specifications
ColorBlack
MaterialNatural Rubber
TypeDouble Blower
Unit SizeEA