Disposable Roller Bottles
Disposable Roller Bottles
Actual product may vary
Fisherbrand
Disposable Roller Bottles
Manufactured from optically clear virgin polystyrene for excellent visibility and tissue culture-treated for optimal cell attachment. Corning™ Disposable Roller Bottles allow Initial scale-up of adherent cells with intermittent exposure of cells to the air and the media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCell Growth AreaClosure TypePackaging
FKS10507142850 cm2Easy Grip40/pk.
CRG430849850 cm2Easy Grip40/pk.
FIS06-415-6850 cm2Easy Grip40/pk.
Catalog number FKS10507142
Price (SGD)
2,887.00
PK
Add to cart
Cell Growth Area:
850 cm2
Closure Type:
Easy Grip
Packaging:
40/pk.
Price (SGD)
2,887.00
PK
Add to cart
  • Bottles are manufactured from virgin polystyrene, caps from high density polyethylene
  • Both resins meet the USP Class VI requirements for plastic containers and closures
  • Treated for optimal cell attachment
  • One piece seamless construction with printed graduations
  • Printed lot numbers to aid in product traceability
  • Sterilized by gamma radiation and nonpyrogenic
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Capacity74.3 oz., 2200mL
Capacity (Metric)2200 mL
Capacity (English)74.3 oz.
Diameter (English)4.6 in.
Diameter (English) Outer Neck1.77 in.
Diameter (Metric) Outer Neck45 mm
MaterialPolystyrene
Packaging40/pk.
SterilitySterile
Cell Growth Area850 cm2
Closure ColorOrange
Surface TreatmentTissue culture treated
GraduatedYes
MouthWide
Height (Metric)271 mm
Height (English)10.66 in.
Closure MaterialHDPE
Closure Size48 mm
Diameter (Metric)117 mm
Closure TypeEasy Grip
Unit SizePK