Polypropylene Cryogenic Storage Box
Polypropylene Cryogenic Storage Box
Actual product may vary
Fisherbrand
Polypropylene Cryogenic Storage Box
Store up to 81 microtube and cryogenic vials in vibrantly colored boxes. Fisherbrand™ Polypropylene Microtube Storage Boxes are designed for quick visual identification and easy access within a compact footprint. Safe for low temperature storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS10500203Natural
FKS10243272Assorted (Blue, Green, Orange, Purple, Yellow)
Catalog number FKS10500203
Price (SGD)
141.00
PK
Add to cart
Color:
Natural
Price (SGD)
141.00
PK
Add to cart
Compact footprint for storage of up to 81 microtube and cryogenic vials.
  • Made from polypropylene
  • Store microtubes or cryogenic vials
  • Easy open, friction fit lid
  • Safe for low temperature storage
  • Tubes: 1.5/2.0mL
  • Wells: 81
  • Well Dia: 12.3mm
  • Autoclavable
Each Fisherbrand™ Microtube Storage box is designed for quick visual identification.
  • Uniform wells with location ID
  • Tube capacity is indicated on the outside of the box
  • Translucent vibrant colors with a frosted labeling area
  • Debossed with a distinctive wave design
Specifications
Closure TypeFriction-Fit Lid
ColorNatural
MaterialPolypropylene
ShapeSquare
TypeCryo Storage Box
For Use With (Application)Microtube Storage
Holds81
Quantity5 Pack
Tube Array9 x 9
Dimensions (L x W x H)130 x 130 x 47 mm
Length (Metric)130 mm
Width (Metric)130 mm
Height (Metric)47 mm
Weight (Metric)0.73 kg
Unit SizePK