H-Foot Stand ST-F 10/600 for Polytron™ PT 3100 D
H-Foot Stand ST-F 10/600 for Polytron™ PT 3100 D
Actual product may vary
Fisherbrand
H-Foot Stand ST-F 10/600 for Polytron™ PT 3100 D
An H-foot stand made of anodized aluminum.
Have Questions?
Catalog number FKS10495342
Price (SGD)
10,396.00
PK
Add to cart
Price (SGD)
10,396.00
PK
Add to cart
  • Height adjustable
  • Swiveling PTFE-coated work table with vessel holder
  • 5.5kg in weight
Specifications
Product TypeHomogenizer Stand
MaterialAnodized Aluminum
DescriptionH-foot homogenizer stand
For Use With (Equipment)ST-F 10/600
Unit SizePK