Check Order Status
Freeze Drying Stoppers
Actual product may vary
Fisherbrand
Freeze Drying Stoppers
Push in to engage and pull out to remove stopper. Azpack™ Freeze Drying Stoppers are nonthreaded and constructed of sturdy rubber.
Have Questions?
Catalog number
FKS10483274
Price (SGD)
542.00
PK
Add to cart
Price (SGD)
542.00
PK
Add to cart
For use with Freeze Drying and Injection Vials
Nonthreaded push-in stopper
Use with appropriate polypropylene screw thread cap
Specifications
Material
Bromobutyl Rubber
Quantity
500 Pack
For Use With (Equipment)
2mL glass vials
Diameter (Metric)
13 mm