Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Closure Size | Base Style | Material |
---|---|---|---|---|
FKS10477614 | 1000 μL | 11 mm | Flat Bottom | Polypropylene |
FIS03-377-299 | 475 μL | 11 mm | Conical Base | Polypropylene |
C4011-11 | 800 μL | 11 mm | Conical Base | Polypropylene |
FIS22-045618 | 800 μL | 11 mm | Conical Base | Polypropylene |
Choose from these chemically inert polypropylene vials, manufactured from high-quality, GC-tested polypropylene material, that are suitable for most chromatography applications. Thermo Scientific™ 11mm Plastic Crimp/Snap Top Vials provide an economical alternative to glass for some applications. Polypropylene is chemically inert and suitable for most chromatography applications, and can be used when levels of inorganic ions have to be kept to an absolute minimum. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.