Microlance™ Stainless Steel Needles
Microlance™ Stainless Steel Needles
Actual product may vary
Fisherbrand
Microlance™ Stainless Steel Needles
Color coded for easy recognition of the needle’s guage. BD™ Microlance™ Stainless Steel Needles feature thin walls for optimal flow and fit Luer slip syringes. These sterile, individually wrapped needles ensure more comfortable insertion with less risk for punching.
Have Questions?
Catalog NumberNeedle GaugeLength (English) NeedleDiameter (Metric) OuterColor
FKS1047220421 G1.5 in.0.8 mmGreen
Catalog number FKS10472204
Price (SGD)
22.00
PK
Add to cart
Needle Gauge:
21 G
Length (English) Needle:
1.5 in.
Diameter (Metric) Outer:
0.8 mm
Color:
Green
Price (SGD)
22.00
PK
Add to cart
Unique slip angle and polishing method results in less patient discomfort at the injection site and less risk for punching.
  • Hypodermic needles with thin walls for optimal flow
  • Fits Luer slip syringes—For use with SZR-150
  • Color coded for quick recognition of gauge for easy selection of appropriate flow rate
  • Sterile and individually wrapped for hygienic, single-use application
  • Stainless steel needles with epoxy resin adhesives
Specifications
MaterialStainless Steel with Epoxy Resin Adhesives
ColorGreen
Description5,000 individually wrapped needles, thin cannula wall
Needle Gauge21 G
Diameter (Metric) Outer0.8 mm
For Use With (Application)For single use only
Latex FreeYes
Length (English) Needle1.5 in.
Product TypeNeedle
Length (Metric) Needle40 mm
Needle Point StyleBevel
For Use With (Equipment)SZR -150, connected to a Luer adapter or Luer Lock connector