Sputum Collection Container
Sputum Collection Container
Actual product may vary
Fisherbrand
Sputum Collection Container
Manufactured with polypropylene for chemical resistance and durability. Kartell™ Sputum Collection Containers are ideal for the storage, collection and transportation of specimens.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1046997230 to 55 mL
Catalog number FKS10469972
Price (SGD)
417.00
PK
Add to cart
Capacity (Metric):
30 to 55 mL
Price (SGD)
417.00
PK
Add to cart
  • Autoclavable
  • Leakproof lid
  • Ideal for stoage, collection and transportation of specimens
  • CE-IVD approved
Specifications
MaterialPolypropylene
Capacity (Metric)30 to 55 mL
Certifications/ComplianceCE and IVD
Closure Size57 mm
Diameter (Metric)Mouth: 52.1 mm, Bottom: 45 mm
Height (Metric)34.1 mm
No. per Pack500/Pk.
Unit SizePK