1x1 image pixel for data collection
Stackable Graduated Beaker with Open Handle
Stackable Graduated Beaker with Open Handle
Actual product may vary
Fisherbrand
Stackable Graduated Beaker with Open Handle
Stack beakers when not in use to save space. Buerkle™ Stackable Graduated Beaker with Open Handle include blue graduation marks for easy volume assessment.
Have Questions?
Catalog NumberCapacity (Metric)
FKS104687111000 mL
Catalog number FKS10468711
Price (SGD)
33.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
33.00
EA
Add to cart
Graduated beaker with graduation printed in blue. Beakers can be stacked to save space during storage.
  • Open handle with integral pour spout
  • Polypropylene
  • Conical shape
  • Transparent
  • Autoclavable
  • Shatterproof
  • Good chemical resistance
Specifications
ColorTranslucent
Diameter (Metric)Top125 mm
Capacity (Metric)1000 mL
MaterialPolypropylene
For Use With (Application)Suitable for Foodstuff
Height (Metric)167 mm
IncludesHandle, Integral Spout
Increments10 mL
QuantityEach
TypeBeaker
AutoclavableAutoclavable
GraduatedYes
ScaleBlue Graduated Scale
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link