Manufactured from robust, lightweight polypropylene for durability and excellent chemical resistance. Kartell™ Polypropylene Buchner Funnels with an integral filter is ideal for separating solids from liquids in a laboratory.
Have Questions?
Catalog Number
Capacity (Metric)
Diameter (Metric)Top
FKS10459592
40 mL
42.5mm
Catalog number FKS10459592
Price (SGD)
35.00
PK
Add to cart
Capacity (Metric):
40 mL
Diameter (Metric)Top:
42.5mm
Price (SGD)
35.00
PK
Add to cart
Manufactured from robust, lightweight polypropylene for durability and excellent chemical resistance. Kartell™ Polypropylene Buchner Funnels with an integral filter is ideal for separating solids from liquids in a laboratory.
Unbreakable, resistant to corrosion and heat
Two piece construction allows top and bottom to be separated for easy clean up
Autoclavable
Suitable for foodstuff
Specifications
Diameter (Metric)Top42.5mm
Capacity (Metric)40 mL
MaterialPolypropylene
AutoclavableAutoclavable
DescriptionBuchner filter
For Use With (Application)Filtration, separating solids from liquids, collecting recrystallized compounds