NucleoVac Vacuum Regulator
NucleoVac Vacuum Regulator
Actual product may vary
Fisherbrand
NucleoVac Vacuum Regulator
Ensures a reliable vacuum source when manually processing DNA columns under vacuum. Macherey-Nagel™ NucleoVac Vacuum Regulator is designed for use with the NucleoVac Vacuum Manifold for fast processing of large sample volumes via parallel processing of columns.
Have Questions?
Catalog NumberQuantity
FKS104595611 Ea.
Catalog number FKS10459561
Price (SGD)
726.00
EA
Add to cart
Quantity:
1 Ea.
Price (SGD)
726.00
EA
Add to cart
Vacuum filtration columns are used to isolate and purify nucleic acids, including plasmid DNA. A reliable vacuum source is critical for successful manual processing. A vacuum regulator is recommended to control the vacuum pressure levels, which impacts the flow rate of the sample through the column. In addition, a regulator controls the vacuum time to ensure complete filtration of the sample.
The NucleoVac Vacuum Regulator is designed for use with the NucleoVac Vacuum Manifold and with various DNA isolation/purification column systems, including NucleoSpin™, NucleoFast™ and NucleoSnap kits.
Specifications
Product TypeNucleoVac Vacuum Regulator
DescriptionFor use with NucleoVac Vacuum Manifold and DNA isolation/purification columns (e.g., NucleoSpin™ NucleoFast™ or NucleoSnap™ kits)
For Use With (Application)For controlling of vacuum
Quantity1 Ea.
Unit SizeEA