Product Image
Quickfit
BCS™ XP System, Complete
The Behring Coagulation System (BCS™ XP System) performs quantitative assays of specific parameters in human citrated plasma.
Have Questions?
Catalog NumberThroughput
FKS10459330380 PT Tests/Hr., 325 Simultaneous PT/APTT Tests/Hr.
Catalog number FKS10459330
Price (SGD)
281.00
Each
Throughput:
380 PT Tests/Hr., 325 Simultaneous PT/APTT Tests/Hr.
Price (SGD)
281.00
Each
The BCS™ XP System, one of the most widely used fully automated hemostasis analyzers, delivers accurate and precise results. The high-speed, random-access analyzer performs clotting, chromogenic, immunologic, and agglutination testing while allowing the consolidation of both specialty and routine reagents onto one high throughput analyzer.
  • User-friendly software and customizable interface allow for versatile user-defined configurations to meet your lab’s specific needs.
  • Automatic monitoring of reagent levels and onboard stability tracking enable personnel to focus on what’s important—critical results.
  • Broad routine and specialty testing menu, including vWF ristocetin and INNOVANCE™ ETP, allow for platform consolidation.
  • Reagent and sample probe crash sensors protect capped reagent bottles or sample tubes from damage.
  • True random-access and walk-away ability for both normal and pathological samples maximize high-volume routine workloads.
Specifications
TypeBCS XP System
Sample Size100 Samples (10 Racks)
Throughput380 PT Tests/Hr., 325 Simultaneous PT/APTT Tests/Hr.
Depth (Metric)630 mm
Height (Metric)920 mm
Width (Metric)1240 mm
Unit SizeEach