Quickfit™ Plain Digestion Tubes
Product Image
Fisherbrand
Quickfit™ Plain Digestion Tubes
For use with block heaters
Have Questions?
Catalog NumberDiameter (Metric) OuterDescription
FKS1045916026 mmCapacity: 100mL
Catalog number FKS10459160
Price (SGD)
561.00
PK
Add to cart
Diameter (Metric) Outer:
26 mm
Description:
Capacity: 100mL
Price (SGD)
561.00
PK
Add to cart
  • With plain beaded neck
  • Manufactured from borosilicate glass for highest chemical and thermal durability
Specifications
DescriptionCapacity: 100mL
MaterialBorosilicate Glass (3.3)
Diameter (Metric) Outer26 mm
Overall Length (Metric)300 mm
Product TypeDigestion Tube
Unit SizePK