Bacto™ Dehydrated Agar
Actual product may vary
Fisherbrand
Bacto™ Dehydrated Agar
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
Have Questions?
Catalog NumberQuantityPackaging Type
FKS10455513454 gBottle
Catalog number FKS10455513
Price (SGD)
971.00
Pack
Quantity:
454 g
Packaging Type:
Bottle
Price (SGD)
971.00
Pack
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling
Determines motility and growth of anaerobes and microaerophiles
Specifications
Certifications/ComplianceISO/DIS, 11014
Concentration1.5%
AdditiveAgar
FormDehydrated
For Use With (Application)Determination of motility and the growth of anaerobes and microaerophiles
Packaging TypeBottle
Quantity454 g
Shelf Life1825 Days
Unit SizePack