Counting Chambers, Neubauer Pattern, BLAUBRAND™, double ruling
Counting Chambers, Neubauer Pattern, BLAUBRAND™, double ruling
Actual product may vary
Fisherbrand
Counting Chambers, Neubauer Pattern, BLAUBRAND™, double ruling
BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.
Have Questions?
Catalog NumberIncludes
FKS10450751With spring clips
Catalog number FKS10450751
Price (SGD)
346.00
PK
Add to cart
Includes:
With spring clips
Price (SGD)
346.00
PK
Add to cart
  • Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
  • The ruling shows 9 large squares of 1 mm2 each.
  • The four large squares in the corners marked "L" for counting leucocytes are each subdivided into 16 squares with 0.25 mm sides.
  • The large square in the center is subdivided into 16 squares with 0.25 mm sides.
  • Each group square consists of 16 mini squares with 0.05 mm sides, each having an area of 0.0025 mm2.
  • The 5 group squares in the large central square are used for counting thrombocytes and erythrocytes.
  • Contrary to the more advanced Improved Neubauer counting chambers, the counting area of each group square is limited by the outer one of the triple boundary lines.
Specifications
Product TypeCounting Device
Quantity1Each
Depth (Metric)0.1mm
Certifications/ComplianceDIN EN 285
For Use With (Application)Counting leucocytes, thrombocytes and erythrocytes
IncludesWith spring clips
Unit SizePK