Polycarbonate Desiccators
Polycarbonate Desiccators
Actual product may vary
Fisherbrand
Polycarbonate Desiccators
Includes polycarbonate lid, polypropylene base and desiccant tray. BRAND™ Polycarbonate Desiccators feature a venting stopper with non-return valve in lid and polychlorobutadiene rubber (CK) O-ring compressed by vacuum to provide the seal between lid and base. Light-weight and easy to handle.
Have Questions?
Catalog NumberDiameter (Metric) OuterHeight (Metric)
FKS10444641170 mm195 mm
Catalog number FKS10444641
Price (SGD)
399.00
EA
Add to cart
Diameter (Metric) Outer:
170 mm
Height (Metric):
195 mm
Price (SGD)
399.00
EA
Add to cart
  • Includes polycarbonate lid, polypropylene base and desiccant tray.
  • Venting stopper with non-return valve in lid.
  • Polychlorobutadiene rubber (CK) O-ring, compressed by vacuum provides seal between lid and base.
  • Light-weight and easy to handle.
Specifications
MaterialPolycarbonate Lid, Polypropylene Base and Dessicant Tray
Product TypeDesiccator
IncludesPolychlorobutadiene rubber (CK) O-ring
Height (Metric)195 mm
Diameter (Metric) Outer170 mm
Unit SizeEA