Automatic Burettes, Acc. to Dr. Schilling
Automatic Burettes, Acc. to Dr. Schilling
Actual product may vary
Fisherbrand
Automatic Burettes, Acc. to Dr. Schilling
Excellent for automatically dispensing measured amounts of chemical solutions in the laboratory. Hirschmann Instruments™ Automatic Burettes, Acc. to Dr. Schilling are available in a range of capacities to fit your needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1044133125 mL
FKS1019592010 mL
FKS1036000150 mL
Catalog number FKS10441331
Price (SGD)
389.00
EA
Add to cart
Capacity (Metric):
25 mL
Price (SGD)
389.00
EA
Add to cart
  • With Schellbach stripe
  • Blue graduation
  • Fast flow burette
Specifications
Capacity (Metric)25 mL
MaterialGlass
Graduations0.1 mL
IncludesLDPE Bottle
ScaleBlue
Tolerance±0.1 mL
TypeSchilling Buret
Certifications/ComplianceDKD Certificate
Unit SizeEA