Disposable Sample Pans
Disposable Sample Pans
Actual product may vary
Fisherbrand
Disposable Sample Pans
Use with Sartorius™ Moisture Balances. Sartorius™ Disposable Aluminum Sample Pans provide easy clean-up when determining moisture content.
Have Questions?
Catalog NumberdimCapacityMetric
FKS10440253
Catalog number FKS10440253
Price (SGD)
245.00
PK
Add to cart
Price (SGD)
245.00
PK
Add to cart
Use with Sartorius™ Moisture Balances. Sartorius™ Disposable Aluminum Sample Pans provide easy clean-up when determining moisture content.
  •  Aluminum
Specifications
Bottom ShapeFlat
DisposableSingle-use
MaterialAluminum
ShapeCircular
TypeBalance Pan
Diameter (Metric)90 mm
Unit SizePK