Hypodermic Needles
Hypodermic Needles
Actual product may vary
Fisherbrand
Hypodermic Needles
Designed for patient comfort in a broad array of sizes. B Braun™ Hypodermic Needles provide a smooth surface with light silicone coating designed to minimize discomfort.
Have Questions?
Catalog NumberNeedle GaugeLength (Metric) NeedleDiameter (Metric) OuterColor
FKS1043888121 G120 mm0.8 mmGreen
Catalog number FKS10438881
Price (SGD)
180.00
PK
Add to cart
Needle Gauge:
21 G
Length (Metric) Needle:
120 mm
Diameter (Metric) Outer:
0.8 mm
Color:
Green
Price (SGD)
180.00
PK
Add to cart
 Unique, highly automated grinding and polishing process and low-friction lubrication is designed to achieve unequaled sharpness and ease of penetration. Transparent hub design provides high visibility to detect flashback.
  • Color-coded hubs conform to ISO 6009 for easy identity
  • Wide range of diameters and lengths
  • Gauges from 18G to 27G
  • Ultra-sharp, tri-beveled stainless steel designed to result in minimal pain
Specifications
MaterialPolypropylene (Hub), Stainless Chromium Nickel Steel
ColorGreen
Needle Gauge21 G
Diameter (Metric) Outer0.8 mm
For Use With (Application)For minimal pain upon puncture, single use
Latex FreeYes
Product TypeNeedle
Length (Metric) Needle120 mm
Needle Point StyleUltra-sharp Long Tri-bevel