Check Order Status
Fisherbrand™ Premium Microscope Slide Box
Actual product may vary
Fisherbrand
Fisherbrand™ Premium Microscope Slide Box
The Fisherbrand™ microscope slide box holds 100 slides. It has a nickel-plated clasp and is ABS plastic foam lined. These boxes are stackable.
Have Questions?
Catalog number
FKS10428241
Price (SGD)
24.00
EA
Add to cart
Price (SGD)
24.00
EA
Add to cart
The Fisherbrand™ microscope slide box holds 100 slides. It has a nickel-plated clasp and is ABS plastic foam lined. These boxes are stackable.
Store and transport 100 slides safely with durable ABS cases
Standard microscope slides numbered to correspond with slide inventory sheet
Grooved slots
Provides safe, dust-free storage
Box lined with foam
Lid closes securely with rust-resistant, nickel-plated, clasp and pin hinge
Dimensions: 208 x 175 x 34 mm
Specifications
Closure Type
Hinged with clasp
Color
Green
Dimensions (L x W x H)
208 x 175 x 34 mm
Height (Metric)
34 mm
Length (Metric)
208 mm
Material
ABS
Quantity
100 Pk.
Shape
Rectangular
Width (Metric)
175 mm