Millex™-FG Syringe Filter Unit, Hydrophobic PTFE, 0.2 μm
Millex™-FG Syringe Filter Unit, Hydrophobic PTFE, 0.2 μm
Actual product may vary
Fisher Catalogue
Millex™-FG Syringe Filter Unit, Hydrophobic PTFE, 0.2 μm
Features a hydrophobic Fluoropore™ (PTFE) membrane. Millex™-FG Syringe Filter, 0.20μm, Hydrophobic PTFE provides broad chemical compatibility and is ideal for filtering samples in organic solvents or to prevent back-streaming of atmospheric moisture in venting applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)ConnectorsVolume (Metric)Quantity
FKS1042155150 mmHB/HB4 L10/Pk.
FISSLFG0501050 mmHB/HB4 L10/Pk.
Catalog number FKS10421551
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
50 mm
Connectors:
HB/HB
Volume (Metric):
4 L
Quantity:
10/Pk.
Protect vacuum sources from contamination and moisture damage
Millex™ syringe filter units with hydrophobic Fluoropore™ or Millipore Express™ PLUS PES membrane are ideal for in-line sterilization of gases and venting sterile containers, and filters with Fluoropore™ membrane can also be used for sterilizing or clarifying organic solutions. There are also specialized filter units to protect hemodialysis transducers from blood and moisture. The 50 mm Millex™ filter units are especially useful for vacuum line protection. All units are bi-directional.
Features & Benefits:
  • Specialized filter units protect hemodialysis transducers from blood and moisture
  • 50 mm Millex™ units ideal for vacuum line protection
  • All Millex™ Hydrophobic PTFE units are bi-directional in flow
Applications:
Sterilizing Gases, Vacuum Line Protection, Venting Sterile Containers, Sterilizing and Clarifying Organic Solutions
Specifications
AutoclavableAutoclavable
ConnectorsHB/HB
DisposableSingle-use
Material (Membrane)PTFE (Hydrophobic)
Pore Size0.2 μm
SterilityNon-sterile
Filtration Area19.6 cm2
For Use With (Application)Filtering vacuum Sources and Tank/Bioreactor Venting with Analytical Sample Preparation
Housing MaterialPolypropylene
Inlet StyleStepped hose barb with Female Luer Slip
Outlet StyleFemale Luer Slip
Quantity10/Pk.
Volume (Metric)4 L
Length (Metric)71 mm
Diameter (Metric)50 mm
Unit SizePK