Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Actual product may vary
Fisherbrand
Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Manufactured with optically clear polystyrene for accurate measurement. Corning™ Stripette™ Individually Wrapped Plastic or Paper/Plastic Disposable Polystyrene Serological Pipets are ideal for sterile tissue culture applications, especially while wearing latex gloves, as the wrap reduces static cling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Color
FKS104202015 mLBlue
FIS07-200-5711 mLYellow
FIS07-200-57410 mLOrange
FKS1008445010 mLOrange
FKS1060615125 mLRed
FIS07-200-57650 mLPurple
CRG4491100 mLBlue
FIS07-200-620100 mLBlue
FKS10255221100 mLBlue
Catalog number FKS10420201
Price (SGD)
321.00
PK
Add to cart
Volume (Metric):
5 mL
Color:
Blue
Price (SGD)
321.00
PK
Add to cart
  • Sterile
  • RNase-/DNase-free
  • Nonpyrogenic
  • Noncytotoxic
  • BSE/TSE-free
  • Exclusive anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Bi-directional graduations provide choice of ascending and descending scales
  • Negative graduations allow additional working volume
  • Three packaging options: Individually wrapped, paper/plastic; Individually wrapped, clear plastic; Bulk packed
  • Volumetric accuracy of ±2% at full capacity
Specifications
Accuracy±2%
Certifications/ComplianceNon-pyrogenic, DNase, RNase-free
MaterialPolystyrene
SterilitySterile
GraduatedYes
Graduations0.1 mL
Length (English)13.7 in.
Negative Graduations2.5 mL
Quantity200/Cs.
Tip StyleRound
Tolerance±2%
Packaging TypeIndividually Wrapped, Paper/Plastic Packaging
ColorBlue
Volume (Metric)5 mL
Length (Metric)34.8 cm
TypeSerological Pipet