RH basic 2 Magnetic Hotplate Stirrer, Ambient to 320°C, Stainless Steel
RH basic 2 Magnetic Hotplate Stirrer, Ambient to 320°C, Stainless Steel
Actual product may vary
Fisherbrand
RH basic 2 Magnetic Hotplate Stirrer, Ambient to 320°C, Stainless Steel
Stirring Range: 100 to 2000 rpm, Shape: Round, Size: 5 in. dia.
Have Questions?
Catalog NumberTop Plate Material
FKS10417792Stainless Steel
Catalog number FKS10417792
Price (SGD)
-
Top Plate Material:
Stainless Steel
  • Economic magnetic stirrer with stainless steel heating plate
  • Fixed safety circuit 400°C
  • Soft-start stirring motor
  • Stirs quantities of up to 10 liters
Specifications
Top Plate MaterialStainless Steel
Voltage230 V
Controller Type0 to 6 scale (speed)
ModelRH basic 2
Size5 in. dia.
Height (Metric)105 mm
Temperature (Metric)Ambient to 320°C
Ambient Temperature5°C to 40°C
Frequency50/60 Hz
Width (Metric)168 mm
Wattage415 W
Weight (English)5.3 lb.
Weight (Metric)2.4 kg
TypeStirring Hotplate
ShapeRound
No. of Positions1
Max. Container Size10 L
Stirring Range100 to 2000 rpm
Unit SizeEA