Check Order Status
18/10 E-Poli Flask Tongs
Actual product may vary
Fisherbrand
18/10 E-Poli Flask Tongs
Lift and move flasks during experimentation using Bochem™ 18/10 E-Poli Flask Tongs. Their points are polyamid coated.
Have Questions?
Catalog Number
Length (Metric) Opening
FKS10410423
115 mm
Catalog number
FKS10410423
Price (SGD)
91.00
EA
Add to cart
Length (Metric) Opening:
115 mm
Price (SGD)
91.00
EA
Add to cart
Points are polyamid coated
Standard Length: 250mm
Stainless Steel
Specifications
Length (Metric) Opening
115 mm
Length (Metric)
250 mm
Material
Stainless Steel (18/10)
Type
Flask Tongs