Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Actual product may vary
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Fisherbrand
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentCertifiedVolume (Metric) WorkingBarcode
FKS10394751MaxiSorpYes350 μLNo Barcode
FKS10468075Immulon 4 HBXNo280 μLNo Barcode
439454MaxiSorpYes350 μLNo Barcode
FIS12-565-135MaxiSorpYes350 μLNo Barcode
442404MaxiSorpNo350 μLNo Barcode
FKS10498802PolySorpNo350 μLNo Barcode
456537MaxiSorpYes350 μLNo Barcode
Catalog number FKS10394751
Price (SGD)
875.00
PK
Add to cart
Surface Treatment:
MaxiSorp
Certified:
Yes
Volume (Metric) Working:
350 μL
Barcode:
No Barcode
Price (SGD)
875.00
PK
Add to cart
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces:
  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
Specifications
BarcodeNo Barcode
Binding PropertyHydrophilic
CertifiedYes
FormatStandard
LidWithout Lid
Detection MethodColorimetric
Array8 x 12
Packaging12 inner bags of 5 each
ColorClear
Product TypeMicroplate
Skirt StylePinchbar
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
Volume (Metric) Working350 μL
SterilityNon-sterile
Surface TreatmentMaxiSorp
Well ShapeFlat Bottom (F-well)
No. per Case60
No. per Pack5
Plate BlockingNone
Quantity60/Cs.
Target MoleculeAntibodies (Glycoproteins)
Well DesignF96
Unit SizePK