PCR Tubes
PCR Tubes
Actual product may vary
Fisherbrand
PCR Tubes
Constructed of extra-thin wall thickness to provide optimal heat transfer and reduced cycle time. Fisherbrand™ PCR Tubes eliminate contamination and sample loss with high-quality raw materials.
Have Questions?
Catalog NumberColor
FKS10385572Clear
Catalog number FKS10385572
Price (SGD)
-
Color:
Clear
  • Reduced cycle time and optimal heat transfer - guaranteed by extra thin, uniform wall thickness of tube / available with flat or domed cap
  • Effortless and secure working - ergonomic cap design allows opening and closing with one hand
  • Made of materials that meets standards of purity
  • Highly resistant to extreme mechanical strain with excellent chemical resistance
  • Optical measurement and unsurpassed clarity
  • Maintain sample integrity - sterile version available (beta irradiation)
  • Sterile handling - PCR tubes are autoclavable
  • Centrifuge specifications:
    • up to 25.000 x g - fixed-angle rotor
    • up to 30.000 x g - swing out rotor
Specifications
Certifications/ComplianceCE, IVD
ColorClear
Volume (Metric)0.2 mL
Quantity1000 Pack
For Use With (Equipment)Centrifuge
MaterialPolypropylene
Closure TypeFlat Cap
AutoclavableAutoclavable
Product TypePCR Tube
Unit SizePK