Polypropylene Pipette Stand
Polypropylene Pipette Stand
Actual product may vary
Fisherbrand
Polypropylene Pipette Stand
Rotary model, with additional rotary tip reset to protect the pipette tips. Suitable for all graduated and one-mark pipettes. Slightly curved stand base rim helps retain dripping liquids.
Have Questions?
Catalog NumberPositions
FKS1038343194
Catalog number FKS10383431
Price (SGD)
259.00
EA
Add to cart
Positions:
94
Price (SGD)
259.00
EA
Add to cart
  • Polypropylene construction
  • Fits 94 pipettes of different sizes
  • Rotary model
  • Additional rotary tip reset protect the pipette tips
  • Suitable for all graduated and one-mark pipettes
  • Slightly curved stand base rim helps retain dripping liquids
Specifications
MaterialPolypropylene
Positions94
Quantity1 Each
Width (Metric)230 mm
Height (Metric)450 mm
Unit SizeEA