PTFE Giant Stir Bar
PTFE Giant Stir Bar
Actual product may vary
Fisherbrand
PTFE Giant Stir Bar
Designed for use in large containers and vials. These giant PTFE-coated magnetic stir bares are autoclavable.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)Shape
FKS10369640159 mm27 mmCylinder
Catalog number FKS10369640
Price (SGD)
112.00
EA
Add to cart
Length (Metric):
159 mm
Diameter (Metric):
27 mm
Shape:
Cylinder
Price (SGD)
112.00
EA
Add to cart
  • Stable, general-purpose magnetic stir bar
  • Designed for use in large containers and vials
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Diameter (Metric)27 mm
Length (Metric)159 mm
Quantity1/Ea.
ShapeCylinder
Unit SizeEA