PIPETMANTM EasyPackTM Tips
PIPETMANTM EasyPackTM Tips
Actual product may vary
Fisher Catalogue
PIPETMANTM EasyPackTM Tips
Ensure optimal performance of PIPETMAN pipettes across the full volume range with PIPETMANTM EasyPackTM Tips.
Have Questions?
Catalog NumberVolume (Metric)FormatQuantity
FKS103671321 to 10 mLBagged200 (4x50)
Catalog number FKS10367132
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1 to 10 mL
Format:
Bagged
Quantity:
200 (4x50)
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips).
Specifications
Volume (Metric)1 to 10 mL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
AutoclavableAutoclavable
Length (English)7.87 in.
Length (Metric)20 cm
Diameter (Metric) Outer.07 in
CompatibilityPIPETMAN Pipettes
FormatBagged
Tip StyleUniversal
Packaging4 Bags of 50 Tips
Quantity200 (4x50)
Product TypePipet Tips
Color CodeLight Blue
DisposableSingle-use
For Use With (Equipment)Gilson Pipetman
GraduatedYes
Product LinePipetman EasyPack
MaterialPolypropylene
Graduations1 mL
Unit SizePK