Check Order Status
Clear Dropping Bottles with Pipette
Actual product may vary
Fisherbrand
Clear Dropping Bottles with Pipette
Soda-lime glass construction. The interchangeable ground joint stopper has an integrated dropping pipette with rubber cap.
Have Questions?
Catalog Number
Capacity (Metric)
FKS10364991
100 mL
Catalog number
FKS10364991
Price (SGD)
84.00
EA
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
84.00
EA
Add to cart
Clear soda-lime glass construction
Includes dropping pipette and rubber cap
Specifications
Capacity (Metric)
100 mL
Color
Clear
Material
Soda Lime Glass
Closure Included
Yes
Height (Metric)
150 mm
Closure Material
Rubber
Diameter (Metric)
55 mm
Closure Type
Ground Joint Stopper with Rubber Cap