Natural Rubber Pipet Filler
Natural Rubber Pipet Filler
Actual product may vary
Fisherbrand
Natural Rubber Pipet Filler
Manufactured from 100% natural rubber. BRAND™ Natural Rubber Pipet Filler eliminates mouth pipetting and handling of hazardous material. Available in universal, standard, or flip model, with 2 or 3-valve options.
Have Questions?
Catalog NumberVolume (Metric)Model
FKS10363341100 mLUniversal
Catalog number FKS10363341
Price (SGD)
51.00
EA
Add to cart
Volume (Metric):
100 mL
Model:
Universal
Price (SGD)
51.00
EA
Add to cart
  • Manufactured from 100% natural rubber.
  • Eliminates mouth pipetting and handling of hazardous material.
  • Available in universal, standard, or flip model, with 2 or 3-valve options
  • 2-valve flip model enables one-handed operation.
Specifications
DescriptionValve A: Release air, Valve S: Filling, Valve E: Delivery
MaterialNatural Rubber
ModelUniversal
Volume (Metric)100 mL
For Use With (Application)Filling pipets up to 100mL
Includes3 valves
Quantity1 Each
Unit SizeEA