ViscoSampler
ViscoSampler
Actual product may vary
Fisherbrand
ViscoSampler
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)Material
FKS10360201300 mL100 cmPolypropylene
Catalog number FKS10360201
Price (SGD)
1,722.00
EA
Add to cart
Capacity (Metric):
300 mL
Length (Metric):
100 cm
Material:
Polypropylene
Price (SGD)
1,722.00
EA
Add to cart
  • Powerful suction for viscous media
  • The media is drawn up and discharged using the suction flask
  • For use with slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc
  • Easy to clean
  • All surfaces are poreless and without crevices
  • Dirt accumulation is prevented
  • Only round screw threads tried and tested in the field of food hygiene are used
  • Disassembly and cleaning are extremely simple
Specifications
Product TypeViscoSampler
MaterialPolypropylene
Capacity (Metric)300 mL
Length (Metric)100 cm
Unit SizeEA