Azlon™ HDPE, Wide-Neck Aspirator Bottle
Azlon™ HDPE, Wide-Neck Aspirator Bottle
Actual product may vary
Fisherbrand
Azlon™ HDPE, Wide-Neck Aspirator Bottle
For Use With: For bench top dispensing of common laboratory liquids and bulk storage or transportation of liquids and powders
Have Questions?
Catalog number FKS10359554
Price (SGD)
2,512.00
EA
Add to cart
Price (SGD)
2,512.00
EA
Add to cart
For Use With: For bench top dispensing of common laboratory liquids and bulk storage or transportation of liquids and powders
Features:
  • Large capacity
  • Wide opening for easy fill
  • Screw cap with water tight rubber seal to prevent leakage and spills
  • Leak-proof stopcock
  • Carry handles for easy lifting
Uses:
  • Bench top dispensing
  • Bulk storage
  • Transportation
  • Liquids 
  • Powders
Specifications
Capacity (English)26 gal.
Capacity (Metric)100 L
Diameter (Metric) Inner Neck240 mm
For Use With (Application)For bench top dispensing of common laboratory liquids and bulk storage or transportation of liquids and powders
Height (Metric) With Closure770 mm
IncludesClosure, Carrying Handles
MaterialHDPE
Neck StyleWide
Product TypeAspirator Bottle
ShapeCylindrical
Unit SizeEA