CERAN™ Hotplate, 50°C to 500°C, Stainless Steel
CERAN™ Hotplate, 50°C to 500°C, Stainless Steel
Actual product may vary
Fisherbrand
CERAN™ Hotplate, 50°C to 500°C, Stainless Steel
Shape: Rectangle, Size: 17 x 5.5 in.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS10358783440 x 100 x 300 mm
Catalog number FKS10358783
Price (SGD)
5,578.00
EA
Add to cart
Dimensions (L x W x H):
440 x 100 x 300 mm
Price (SGD)
5,578.00
EA
Add to cart
  • The low-mass plate is electrically heated over its full surface and is embedded into a high-grade stainless steel frame which is mounted on an internally insulated stainless steel housing
  • Temperature control system is regulated by an electronic NiCr-Ni temperature sensor with a regulation range between 50° and 500°C
  • Controller is built into a pressure-cast aluminum casing
  • Heating time for 500°C is approx. 8 minutes
  • Four feet, which can be individually adjusted to an approx. height of 8 mm, ensure horizontal stability
  • Equipped with additional sockets for 4 screw-in retort rods (situated 100 mm from each other)
Specifications
Body MaterialStainless Steel
Top Plate MaterialStainless Steel
Size17 x 5.5 in.
Temperature (Metric)50°C to 500°C
Weight (English)11 lb.
Height (Metric)300 mm
Length (Metric)440 mm
Weight (Metric)5 kg
Width (Metric)100 mm
Wattage1500 W
ShapeRectangle
Voltage230 V
TypeHot Plate
Dimensions (L x W x H)440 x 100 x 300 mm
Unit SizeEA