KingFisher™ Plastics for 96 standard and PCR formats
KingFisher™ Plastics for 96 standard and PCR formats
Actual product may vary
Fisherbrand
KingFisher™ Plastics for 96 standard and PCR formats
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Catalog NumberFor Use With (Equipment)Quantity
FKS10357939KingFisher Flex and Presto48/Cs.
Catalog number FKS10357939
Price (SGD)
923.00
EA
Add to cart
For Use With (Equipment):
KingFisher Flex and Presto
Quantity:
48/Cs.
Price (SGD)
923.00
EA
Add to cart
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Format96 Deep-well Plate
Capacity96 Samples
DescriptionKingFisher 96 microplate (200μL) (for Flex and Presto)
*Note: Only works with KingFisher™ Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher™ Flex 96 Deep-Well Heating Block (Cat. No. 24075430).
For Use With (Equipment)KingFisher Flex and Presto
Quantity48/Cs.
Unit SizeEA