PCR Box/Rack
PCR Box/Rack
Actual product may vary
Fisherbrand
PCR Box/Rack
Holds 96-well PCR plates, tube strips, and individual 0.2 mL tubes for easy handling.
Have Questions?
Catalog number FKS10352543
Price (SGD)
219.00
PK
Add to cart
Price (SGD)
219.00
PK
Add to cart
  • Designed to securely accommodate 96-well PCR plates, tube strips, and individual 0.2 mL tubes for effortless handling
  • Alphanumerically marked for convenient sample identification
  • Stackable design (without lids) for efficient storage
  • Manufactured from study, chemically resistant polypropylene
  • Withstands temperatures from -80°C to +121°C
  • Available in a pack of five assorted colors: green, red, violet, yellow, and blue
Specifications
AutoclavableAutoclavable
TypeBox/Rack
MaterialPolypropylene
Height (Metric)4.14 cm
Width (Metric)9.52 cm
Unit SizePK