MF-Millipore™ Membrane Filter, 1.2 μm
MF-Millipore™ Membrane Filter, 1.2 μm
Actual product may vary
Fisherbrand
MF-Millipore™ Membrane Filter, 1.2 μm
Mixed cellulose esters (MCE) membrane, hydrophilic
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS1034018147 mm
FKS1029820025 mm
Catalog number FKS10340181
Price (SGD)
877.00
PK
Add to cart
Diameter (Metric):
47 mm
Price (SGD)
877.00
PK
Add to cart
Used in analytical research.
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
AutoclavableNot Autoclavable
ColorWhite
Filter TypeMembrane Filter
MaterialMCE
Pore Size1.2 μm
SterilityNon-sterile
Flow Rate270 mL/min x cm2
HydrophobicityHydrophilic
Membrane TypePlain
No. per Case100Pack
Quantity100/Pk.
TypeFilter Membranes
WettabilityHydrophilic
Diameter (Metric)47 mm
Unit SizePK