Stand
Stand
Actual product may vary
Fisherbrand
Stand
Have Questions?
Catalog number FKS10328811
Price (SGD)
272.00
EA
Add to cart
Price (SGD)
272.00
EA
Add to cart
VACUBOY hand operator set comprises of: VACUBOY hand operator, stand, rubber adapter for Pasteur pipettes, 1 channel stainless steel aspiration adapter 40mm, 8 channel stainless steel aspiration adapter 40mm, 1 channel plastic aspiration adapter for disposable tips, 1 channel plastic aspiration adapter for disposable tips with ejector, 8 channel plastic aspiration adapter for disposable tips with ejector. Can be used with VACUSAFE comfort vacuum system. Full range of accessories available. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
  • All parts in contact with fluids are autoclavable
Specifications
DescriptionVacuboy bench stand
IncludesDoes not include aspirator
Unit SizeEA