1x1 image pixel for data collection
Orion™ Single Junction Combination pH Electrodes
Orion™ Single Junction Combination pH Electrodes
Actual product may vary
Orion™ Single Junction Combination pH Electrodes
Orion™ Single Junction Combination pH Electrodes
Orion™ Single Junction Combination pH Electrodes
Orion™ Single Junction Combination pH Electrodes
Fisherbrand
Orion™ Single Junction Combination pH Electrodes
Conduct routine, small volume or flat surface pH measurement with Thermo Scientific™ Orion™ economy series pH electrodes.
Have Questions?
Catalog NumberConnector TypesTypeTip Type
FKS10314191BNCCombination pH ElectrodeSemi-micro
Catalog number FKS10314191
Price (SGD)
1,038.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Connector Types:
BNC
Type:
Combination pH Electrode
Tip Type:
Semi-micro
Price (SGD)
1,038.00
EA
Add to cart
Thermo Scientific™ Orion™ Low Maintenance pH Electrodes provide reliable and accurate measurements of routine samples. 9106BNWP is best suited for field or plant use with general purpose samples, and educational institutes where minimal maintenance is required. 911600 is ideal for samples as small as 0.2mL. 912600 is suited for use with long neck flask. 913600 has a flat sensing bulb good for measurement of flat, soft, moist, semi-solid or small volume samples.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required 
Electrode storage solution
910001     pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Warranty period
3 months from date of purchase
Specifications
DescriptionPermanently gelled Ag/AgCl electrolyte
Connector TypesBNC
JunctionWick
MaterialEpoxy
pH Range0 to 12
Temperature (Metric) Operating0°C to 80°C
TypeCombination pH Electrode
For Use With (Equipment)Orion pH Meter
Reference ElectrolyteAg/AgCl
Temperature (English) Operating32°F to 176°F
Tip TypeSemi-micro
Length (English)5.93 in.
Length (Metric)15 cm
Length (Metric) Cable1 m
Diameter (English) Body0.25 in.
Diameter (English) Cap0.18 in.
Diameter (Metric) Body6 mm
Diameter (Metric) Cap10 mm
RefillableNon-refillable
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection