Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Actual product may vary
Fisherbrand
Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Consisting of a membrane-containing filter unit within a polypropylene centrifuge tube. Corning™ Costar™ Spin-X™Centrifuge Tube Filters effectively filters small sample volumes by centrifugation for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and purification of DNA from agarose and polyacrylamide gels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeSterility
FKS103103610.22 μmSterile
FIS07-200-3850.22 μmSterile
FIS07-200-3870.45 μmSterile
Catalog number FKS10310361
Price (SGD)
1,336.00
PK
Add to cart
Pore Size:
0.22 μm
Sterility:
Sterile
Price (SGD)
1,336.00
PK
Add to cart
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Specifications
Material (Membrane)Cellulose Acetate
Pore Size0.22 μm
SterilitySterile
TypeSpin-X
Volume (Metric)0.5 mL
Unit SizePK