L/S™ Platinum-Cured Silicone Precision Tubing
L/S™ Platinum-Cured Silicone Precision Tubing
Actual product may vary
Masterflex
L/S™ Platinum-Cured Silicone Precision Tubing
Ensure optimal performance from your Masterflex™ pump with this silicone pump tubing. Masterflex™ L/S™ Platinum-Cured Silicone Precision Tubing provides accurate, repeatable flow. Ideal for Pharmaceutical, biotech, peristaltic pumping and food applications.
Have Questions?
Catalog NumberTubing Size
FKS1029488216
Catalog number FKS10294882
Price (SGD)
585.00
Each
Add to cart
Tubing Size:
16
Price (SGD)
585.00
Each
Add to cart
Ensure optimal performance from your Masterflex™ pump with this silicone pump tubing. Masterflex™ L/S™ Platinum-Cured Silicone Precision Tubing provides accurate, repeatable flow. Ideal for Pharmaceutical, biotech, peristaltic pumping and food applications.
Masterflex™ L/S™ pump tubing is manufactured to extremely close tolerances that match our L/S™ pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.
  • Fewer potential leachables
  • Excellent biocompatibility
  • Odorless and nontoxic, with no taste imparted to transported fluids
  • Resistant to fungus growth, weather, ozone, corona and radiation
  • Smoother surface than peroxide-cured silicone for reduced protein binding
  • Excellent heat resistance, low compression set, odorless, nontoxic
  • Translucent
Specifications
Certifications/ComplianceUSP Class VIFDA 21 CFR 177.2600 Exceeds 3 A sanitary standards European Pharmacopoeia (EP)
AutoclavableAutoclavable
Diameter (English) Inner3/25 in.
Diameter (Metric) Inner3.1 mm
MaterialSilicone
Diameter (English) Outer0.62 in.
Diameter (Metric) Outer16 mm
Overall Length (English)25 ft.
Overall Length (Metric)760 cm
For Use With (Application)Pharmaceutical, Biotechnology, Peristaltic Pumping and Food Applications
Min. Operating Temperature (English)-58°F
Min. Operating Temperature (Metric)-50°C
Tubing Size16
Flow Rate0.8 to 480 mL/min
Hose Barb0.12 in.
Sterilization MethodAutoclave, Ethylene Oxide, Gamma Irradiation
Height (Metric)8.8 m Water
Height (English)29 ft. Water
Gas PermeabilityCO2: 25,147, O2: 4715, N2: 2284
ColorTranslucent
Max. Operating Temperature (English)446°F
Max. Operating Temperature (Metric)230°C
Product TypePump Tubing
Unit SizeEach