Short-Term Detector Tubes - Hydrogen Sulfide
Short-Term Detector Tubes - Hydrogen Sulfide
Actual product may vary
Fisherbrand
Short-Term Detector Tubes - Hydrogen Sulfide
Have Questions?
Catalog NumberComponents MeasuredMeasurement Range
FKS10276821Hydrogen Sulfide 100/a100 to 2000ppm
Catalog number FKS10276821
Price (SGD)
534.00
PK
Add to cart
Components Measured:
Hydrogen Sulfide 100/a
Measurement Range:
100 to 2000ppm
Price (SGD)
534.00
PK
Add to cart
Specifications
Components MeasuredHydrogen Sulfide 100/a
Humidity Range3 to 40mg/L
Measurement Range100 to 2000ppm
No. of Strokes1
Quantity10
Response Time30 sec.
Temperature Range (English)32°F to 104°F
Temperature Range (Metric)0°C to 40°C
Unit SizePK