Check Order Status
Borosilicate Glass Conical Ground Joint Stopper
Actual product may vary
Fisherbrand
Borosilicate Glass Conical Ground Joint Stopper
Solid or semi-hollow with octagonal grip. BRAND™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3. to DIN 12252 standards.
Have Questions?
Catalog Number
Standard Taper Joint
FKS10273821
NS 14/23
Catalog number
FKS10273821
Price (SGD)
39.00
EA
Add to cart
Standard Taper Joint:
NS 14/23
Price (SGD)
39.00
EA
Add to cart
Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
Solid or semi-hollow
Octagonal grip
Specifications
Quantity
1/Pk.
Standard Taper Joint
NS 14/23
Material
Borosilicate Glass
Product Type
Ground Joint Stopper
Shape
Octagonal Grip (Solid)