Whatman™ 1PS Disposable Phase Separating Paper
Whatman™ 1PS Disposable Phase Separating Paper
Actual product may vary
Fisher Catalogue
Whatman™ 1PS Disposable Phase Separating Paper
Creates a hydrophobic filter that can replace separatory funnel in some applications. Whatman™ 1PS Disposable Phase Separating Paper is impregnated with stable silicone to allow solvent phases to pass through while retaining aqueous phases.
Have Questions?
Catalog NumberDiameter (Metric) Inside
FKS10272081150 mm
Catalog number FKS10272081
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Inside:
150 mm
Creates a hydrophobic filter that can replace separatory funnel in some applications. Whatman™ 1PS Disposable Phase Separating Paper is impregnated with stable silicone to allow solvent phases to pass through while retaining aqueous phases.
Passes organic solvent phases lighter or heavier than water but retains water phase.
  • Hydrophobic
  • Use folded in conical funnel
  • Disposable
  • Eliminates need for washing and drying separatory funnels, cross-contamination
Specifications
MaterialFiberglass/Silicone
For Use With (Application)Liquid phase separation
No. per Pack100Pack
TypeSeparator Paper
Volume (English)2.53 oz.
Volume (Metric)75 mL
Diameter (English) Inside5.9 in.
Diameter (Metric) Inside150 mm
Unit SizePK