Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C
Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C
Actual product may vary
Fisherbrand
Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C
Combines fine particle retention with good flow rate. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C are ideal for the collection of suspended solids in potable water, natural and industrial wastes.
Have Questions?
Catalog NumberDiameter (Metric)PackagingQuantity
FKS1026160170 mmBulk100 Pieces
Catalog number FKS10261601
Price (SGD)
479.00
PK
Add to cart
Diameter (Metric):
70 mm
Packaging:
Bulk
Quantity:
100 Pieces
Price (SGD)
479.00
PK
Add to cart
Combines fine particle retention with good flow rate. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C are ideal for the collection of suspended solids in potable water, natural and industrial wastes.
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500°C. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.
Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
  • Medium to fast, high loading
  • Airflow: 6.7s/100mL
  • Air Monitoring
  • Clarification of aqueous liquids containing low to medium levels of fine particulates
  • Cell harvesting
  • Liquid scintillation counting
  • Marine chemistry and biochemistry
  • Radioimmunoassay procedures
Specifications
Basis Weight53 g/m2
DescriptionGE’s Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis.
FormatDisc
MaterialBorosilicate Glass
Particle Retention1.2 μm
Flow Rate10.5 sec./100 mL
GradeGrade GF/C
PackagingBulk
Quantity100 Pieces
ShapeCircle
Temperature (English) Max.932°F
Temperature (Metric) Max500°C
Thickness (English)0.01 in.
Thickness (Metric)260 μm
TypeFilter
Waterflow Rate105 mL/min
Diameter (English)2.7 in.
Diameter (Metric)70 mm
Unit SizePK