Multi Extraction Mantles - 3 Recess Model, Heating Only
Multi Extraction Mantles - 3 Recess Model, Heating Only
Actual product may vary
Multi Extraction Mantles - 3 Recess Model, Heating Only
Multi Extraction Mantles - 3 Recess Model, Heating Only
Fisherbrand
Multi Extraction Mantles - 3 Recess Model, Heating Only
Vented case's unique airflow ensures the unit remains cool-to-the-touch
Have Questions?
Catalog number FKS10259858
Price (SGD)
5,518.00
EA
Add to cart
Price (SGD)
5,518.00
EA
Add to cart
Vented case's unique airflow ensures the unit remains cool-to-the-touch
  • Individual built-in solid state electronic controls enable easy regulation of each heater, while removing sparking associated with mechanical switching
  • Three 12.7mm (0.5 in.) diameter support rods are included
  • Top cover is polypropylene and external surfaces are powder coated and resistant to most chemical solutions
  • Coiled heating element is suspended within a thermal insulating cartridge to provide maximum heat transfer and support, while removing sparking associated with mechanical switching
  • Individual heater cartridges are replaceable
  • Earth (ground) screen encloses the heaters for added safety
  • Neon lights indicate when power is on and supplied to heaters and, if applicable, supplied to stirrers
Specifications
Certifications/ComplianceCE
Capacity500mL (16.9 oz.)
DesignPolypropylene housing
Capacity (Metric)500 mL
Depth (English)10.25 in.
Depth (Metric)26 cm
For Use WithThree 500 mL Flasks
Height (English)3.5 in.
Height (Metric)8.9 cm
Voltage230 V
Electrical Requirements230V 50/60Hz, 600w (UK and EU plug)
ModelEME30500/CEB
Wattage600w
Width (English)24.8 in.
Width (Metric)63cm
Product TypeHeating Mantle
Temperature (Metric)Ambient to 450°C
Amperage2.6 A
Capacity (English)17 oz.
For Use With (Application)Heating solutions
Unit SizeEA